missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malathion, TRC
Specifications
Specifications
| Chemical Name or Material | Malathion |
| CAS | 121-75-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H19 O6 P S2 |
| Quantity | 5 g |
| Synonym | Butanedioic acid, [(dimethoxyphosphinothioyl)thio]-, diethyl ester (9CI), Succinic acid, mercapto-, diethyl ester, S-ester with O,O-dimethyl phosphorodithioate (6CI,8CI), 8059HC, American Cyanamid 4,049, Atrapa, Carbetovur, Carbetox, Carbofos, Carbophos, Cimexan, Compound 4049, Cythion, Derbac M, Diethyl mercaptosuccinate S-ester with O,O-dimethyl phosphorodithioate, ENT 17,034, ENT 17034, Ethiolacar, Etiol, Extermathion, Forthion, Fosfothion, Fosfotion, Fyfanon, Gammaxine, Hi-Yield, Hilthion, Hymal, IFO 13140, Insecticide 4049, Insecticide no. 4049, Karbofos, Kunamala, Malafor, Malamar, Malamar 50, Malasol, Malaspray, Malataf, Malataf 57EC, Malathine, Malathion, Malathion E 50, Malathion LV Concentrate, Malathion ULV, Malathyl, Malathyne, Malatol, Malatol 500CE, Maldison, Mavidan, Mercaptothion, Moscarda, NSC 6524, Nassrlathion, O,O-Dimethyl S-(1,2-dicarbethoxyethyl) dithiophosphate, Oleophosphothion, Organoderm, Ortho Malathion, Ovide, Phosphothion, Prioderm, Radotion, Radotion E 50, Radotion P 5, S-[1,2-Bis(carbethoxy)ethyl] O,O-dimethyl dithiophosphate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl phosphorodithioate, S-[1,2-Bis(ethoxycarbonyl)ethyl] O,O-dimethyl thiophosphate, SF 60, Sadofos, Sadophos, Security, Siptox I, Suleo M, Sumitox, TAK, TAK (pesticide), TM 4049, Vetiol, Zithiol, [(Dimethoxyphosphinothioyl)thio]butanedioic acid diethyl ester |
| InChI Formula | InChI=1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3 |
| SMILES | CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC |
| IUPAC Name | diethyl 2-dimethoxyphosphinothioylsulfanylbutanedioate |
| Molecular Weight (g/mol) | 330.36 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?