missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Lisofylline, TRC

Product Code. 30482732
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482732 10 mg 10mg
30476677 25 mg 25mg
30476235 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482732 Supplier Toronto Research Chemicals Supplier No. TRCL46905010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6493-06-7
Molecular Formula C13 H20 N4 O3
Molecular Weight (g/mol) 280.32
Synonym 1H-Purine-2,6-dione, 3,7-dihydro-1-(5-hydroxyhexyl)-3,7-dimethyl-, Theobromine, 1-(5-hydroxyhexyl)- (7CI,8CI), 3,7-Dihydro-1-(5-hydroxyhexyl)-3,7-dimethyl-1H-purine-2,6-dione, (±)-1-(5-Hydroxyhexyl)-3,7-dimethylxanthine, (±)-Lisofylline, (±)-Lisophylline, 1-(5-Hydroxyhexyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione, 1-(5-Hydroxyhexyl)-3,7-dimethylxanthine, 1-(5-Hydroxyhexyl)-3,7-dimethylxantine, 1-(5'-Hydroxyhexyl)-3,7-dimethylxanthine, 3,7-Dimethyl-1-(5-hydroxyhexyl)xanthine, BL 194, Penthydroxyfylline, Pentoxifylline alcohol
IUPAC Name 1-(5-hydroxyhexyl)-3,7-dimethylpurine-2,6-dione
SMILES CC(O)CCCCN1C(=O)N(C)c2ncn(C)c2C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H20N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8-9,18H,4-7H2,1-3H3
Formula Weight 280.1535
Chemical Name or Material (+/-)-Lisofylline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.