missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Vortioxetine, TRC

Product Code. 30483305
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483305 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483305 Supplier Toronto Research Chemicals Supplier No. TRCN620560100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitrosovortioxetine
CAS 2925447-38-5
Purity Grade Notes HPLC
Molecular Formula C18 H21 N3 O S
Quantity 100 mg
Synonym N-Nitrosovortioxetine
InChI Formula InChI=1S/C18H21N3OS/c1-14-7-8-17(15(2)13-14)23-18-6-4-3-5-16(18)20-9-11-21(19-22)12-10-20/h3-8,13H,9-12H2,1-2H3
SMILES Cc1ccc(Sc2ccccc2N3CCN(CC3)N=O)c(C)c1
IUPAC Name 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]-4-nitrosopiperazine
Molecular Weight (g/mol) 327.4438
Formula Weight 327.1405
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.