missing translation for 'onlineSavingsMsg'
Learn More

Methyl N-Methoxy-2-methylphenylcarbamate, TRC

Product Code. 30483772
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483772 Supplier Toronto Research Chemicals Supplier No. TRCM2587251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Methyl N-Methoxy-2-methylphenylcarbamate
CAS 151827-82-6
Molecular Formula C10H13NO3
Quantity 1 g
Synonym Methyl N-Methoxy-N-2-methylphenylcarbamate
InChI Formula InChI=1S/C10H13NO3/c1-8-6-4-5-7-9(8)11(14-3)10(12)13-2/h4-7H,1-3H3
SMILES CON(C(=O)OC)c1ccccc1C
Molecular Weight (g/mol) 195.22
Formula Weight 195.0895
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.