missing translation for 'onlineSavingsMsg'
Learn More

Nojirimycin-1-Sulfonic Acid, TRC

Product Code. 30483811
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483811 50 mg 50mg
30471602 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483811 Supplier Toronto Research Chemicals Supplier No. TRCN63500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114417-84-4
Molecular Formula C6 H13 N O8 S
Molecular Weight (g/mol) 259.23
IUPAC Name [(3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)piperidin-2-yl] hydrogen sulfate
SMILES OC[C@H]1NC(OS(=O)(=O)O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H13NO8S/c8-1-2-3(9)4(10)5(11)6(7-2)15-16(12,13)14/h2-11H,1H2,(H,12,13,14)/t2-,3-,4+,5-,6?/m1/s1
Formula Weight 259.0362
Chemical Name or Material Nojirimycin-1-Sulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.