missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso Azithromycin, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Azithromycin |
| Analyte or Component Names | N-Nitroso Azithromycin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H69 N3 O13 |
| Synonym | (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,8,10,12,14-hexamethyl-6-nitroso-1-oxa-6-azacyclopentadecan-15-one |
| InChI Formula | InChI=1S/C37H69N3O13/c1-14-26-37(10,46)30(42)23(6)40(38-47)18-19(2)16-35(8,45)32(53-34-28(41)25(39(11)12)15-20(3)49-34)21(4)29(22(5)33(44)51-26)52-27-17-36(9,48-13)31(43)24(7)50-27/h19-32,34,41-43,45-46H,14-18H2,1-13H3/t19-,20-,21+,22-,23-,24+,25+,26-,27+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1 |
| Solution Type | Neat |
| SMILES | CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(N=O)[C@H](C)[C@@H](O)[C@]1(C)O |
| IUPAC Name | (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,8,10,12,14-hexamethyl-6-nitroso-1-oxa-6-azacyclopentadecan-15-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?