missing translation for 'onlineSavingsMsg'
Learn More

BRAF35 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30497532 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30497532 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30497532 Supplier Novus Biologicals Supplier No. NBP337920MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRAF35 Polyclonal antibody specifically detects BRAF35 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRAF35
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias BRAF35Sox-like transcriptional factor, high-mobility group 20B, HMG domain-containing protein 2, HMG domain-containing protein HMGX2, HMGX2FLJ26127, HMGXB2member 1-related, SMARCE1R, SMARCE1-related protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human BRAF35 (NP_006330.2).,, Sequence:, MSHGPKQPGAAAAPAGGKAPGQHGGFVVTVKQERGEGPRAGEKGSHEEEPVKKRGWPKGKKRKKILPNGPKAPVTGYVRFLNERREQIRTRHPDLPFPEITKMLGAEWSKLQPTEKQRYLDEAEREKQQYMKELRAYQQSEAYKMCTEKIQEKKIKKEDSSSGLMNTLLNGHKGGDCDGFSTFDVPIFTEEFLDQNKAREAELRRLRKMNVAFEEQNAVLQRHTQSMSSARERLEQELALEERRTLALQQQLQAVRQALTASFASLPVPGTGETPTLGTLDFYMARLHGAIERDPAQHEKLIVRIKEILAQVASEHL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10362
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.