Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ASTM Certified, Pore Sizes: 10mm and Larger
Brand: RETSCH 60.159.090000
Additional Details : Weight : 1.00000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
40mm |
For separation, fractioning, particle size determination | |
305 mm | |
90mm | |
ASTM E 11 | |
Shaker AS 300 |