missing translation for 'onlineSavingsMsg'
Learn More

NDP52 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30512380 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30512380 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30512380 Supplier Novus Biologicals Supplier No. NBP338417AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDP52 Polyclonal antibody specifically detects NDP52 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDP52
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human NDP52 (NP_005822.1).,, Sequence:, TTQGEVEEIEQHNKELCKENQELKDSCISLQKQNSDMQAELQKKQEELETLQSINKKLELKVKEQKDYWETELLQLKEQNQKMSSENEKMGIRVDQLQAQLSTQEKEMEKLVQGDQDKTEQLEQLKKENDHLFLSLTEQRKDQKKLEQTVEQMKQNETTAMKKQQELMDENFDLSKRLSENEIICNALQRQKERLEGENDLLKRENSRLLSYMGLDFNSLPYQVPTSDEGGARQNPGLAYGNPYSGIQESSSPSPLSIKKCPICKADDICDHTLEQQQMQPLCFNCPICDKIFPATEKQIFEDHVFCHSL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10241
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.