missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EEN Polyclonal antibody specifically detects EEN in Human,Monkey,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | EEN |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | CNSA1endophilin-2, EEN fusion partner of MLL, EENSH3 domain-containing GRB2-like protein 1, Endophilin-2, extra 11-19 leukemia fusion, Extra eleven-nineteen leukemia fusion gene protein, MGC111371, SH3 domain protein 2B, SH3-containing Grb-2-like 1 protein, SH3D2Bendophilin-A2, SH3-domain GRB2-like 1, SH3P8 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 219-368 of human EEN (NP_003016.1).,, Sequence:, VDAQLDYHRQAVQILDELAEKLKRRMREASSRPKREYKPKPREPFDLGEPEQSNGGFPCTTAPKIAASSSFRSSDKPIRTPSRSMPPLDQPSCKALYDFEPENDGELGFHEGDVITLTNQIDENWYEGMLDGQSGFFPLSYVEVLVPLPQ |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?