missing translation for 'onlineSavingsMsg'
Learn More

DARS2 Antibody [FITC], Novus Biologicals Biologicals™

Product Code. 30513800 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30513800 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30513800 Supplier Novus Biologicals Supplier No. NBP338469F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DARS2 Polyclonal antibody specifically detects DARS2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DARS2
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias aspartate tRNA ligase 2, mitochondrial, Aspartate--tRNA ligase, aspartyl-tRNA synthetase 2, mitochondrial, aspartyl-tRNA synthetase, mitochondrial, AspRS, ASPRS. LBSL, EC 6.1.1, EC 6.1.1.12, FLJ10514, LBSL, MT-ASPRS, RP3-383J4.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 406-645 of human DARS2 (NP_060592.2).,, Sequence:, ANRNWNSPVANFIMESQRLELIRLMETQEEDVVLLTAGEHNKACSLLGKLRLECADLLETRGVVLRDPTLFSFLWVVDFPLFLPKEENPRELESAHHPFTAPHPSDIHLLYTEPKKARSQHYDLVLNGNEIGGGSIRIHNAELQRYILATLLKEDVKMLSHLLQALDYGAPPHGGIALGLDRLICLVTGSPSIRDVIAFPKSFRGHDLMSNTPDSVPPEELKPYHIRVSKPTDSKAERAH
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55157
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.