missing translation for 'onlineSavingsMsg'
Läs mer

ALG2 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Produktkod. 30513915 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
0.1 mL
Förpackningsstorlek:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
30513915 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30513915 Leverantör Novus Biologicals Leverantörsnummer NBP338473APC

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

ALG2 Polyclonal antibody specifically detects ALG2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen ALG2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias alpha-1,3-mannosyltransferase ALG2, alpha-1,3-mannosyltransferase), asparagine-linked glycosylation 2 homolog (S. cerevisiae, asparagine-linked glycosylation 2 homolog (yeast, asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 2 homolog, CDGII, EC 2.4.1, EC 2.4.1.-, FLJ14511, GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase, hALPG2, homolog of yeast ALG2, NET38
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 197-416 of human ALG2 (NP_149078.1).,, Sequence:, DVLYPSLNVTSFDSVVPEKLDDLVPKGKKFLLLSINRYERKKNLTLALEALVQLRGRLTSQDWERVHLIVAGGYDERVLENVEHYQELKKMVQQSDLGQYVTFLRSFSDKQKISLLHSCTCVLYTPSNEHFGIVPLEAMYMQCPVIAVNSGGPLESIDHSVTGFLCEPDPVHFSEAIEKFIREPSLKATMGLAGRARVKEKFSPEAFTEQLYRYVTKLLV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85365
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.