missing translation for 'onlineSavingsMsg'
Learn More

eIF3K Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30517982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30517982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30517982 Supplier Novus Biologicals Supplier No. NBP338633PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

eIF3K Polyclonal antibody specifically detects eIF3K in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3K
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human eIF3K (NP_037366.1).,, Sequence:, MAMFEQMRANVGKLLKGIDRYNPENLATLERYVETQAKENAYDLEANLAVLKLYQFNPAFFQTTVTAQILLKALTNLPHTDFTLCKCMIDQAHQEERPIRQILYLGDLLETCHFQAFWQALDENMDLLEGITGFEDSVRKFICHVVGITYQHIDRWLLAEMLGDLSDSQLKVWMSKYGWSADESGQIFICSQEESIKPKNIVEKIDFDSVSSIMASSQ
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27335
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.