missing translation for 'onlineSavingsMsg'
Learn More

EEF1D Antibody (0H1) [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30519372 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519372 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519372 Supplier Novus Biologicals Supplier No. NBP342963AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EEF1D Monoclonal antibody specifically detects EEF1D in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1D
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 0H1
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Antigen NY-CO-4, EF-1D, EF-1-delta, EF1DFP1047, elongation factor 1-delta, eukaryotic translation elongation factor 1 delta (guanine nucleotide exchangeprotein), FLJ20897, guanine nucleotide exchange protein
Host Species Mouse
Immunogen eukaryotic translation elongation factor 1 delta(guanine nucleotide exchange protein) - (Uniprot# P29692)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1936
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.