missing translation for 'onlineSavingsMsg'
Learn More

WTAP Antibody (9D4) [Alexa Fluor« 488], Novus Biologicals Biologicals™

Product Code. 30519759 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519759 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519759 Supplier Novus Biologicals Supplier No. NBP343062AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WTAP Monoclonal antibody specifically detects WTAP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WTAP
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9D4
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms' tumour 1-associating protein, WT1-associated protein
Host Species Mouse
Immunogen Wilms tumor 1 associated protein - (Uniprot# Q15007)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9589
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.