missing translation for 'onlineSavingsMsg'
Learn More

FUT9 Antibody (2B6) [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30520151 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520151 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520151 Supplier Novus Biologicals Supplier No. NBP342972UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FUT9 Monoclonal antibody specifically detects FUT9 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT9
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2B6
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, Fucosyltransferase 9, fucosyltransferase 9 (alpha (1,3) fucosyltransferase), Fucosyltransferase IX, fucT-IX, Fuc-TIXFucT-IX, Galactoside 3-L-fucosyltransferase
Host Species Mouse
Immunogen fucosyltransferase 9(alpha(1,3) fucosyltransferase) - (Uniprot# Q9Y231)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10690
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.