missing translation for 'onlineSavingsMsg'
Learn More

RAC2 Antibody (8F0) [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30521681 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30521681 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30521681 Supplier Novus Biologicals Supplier No. NBP343023UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAC2 Monoclonal antibody specifically detects RAC2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAC2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8F0
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Gx, HSPC022, ras-related C3 botulinum toxin substrate 2 (rho family, small GTP bindingprotein Rac2), rho family, small GTP binding protein Rac2, small GTP-bindingprotein Rac2)
Host Species Mouse
Immunogen ras-related C3 botulinum toxin substrate 2(rho family, small GTP binding protein Rac2) - (Uniprot# P15153)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5880
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.