missing translation for 'onlineSavingsMsg'
Learn More

HE4/WFDC2 Antibody (CL12917) [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30528055 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30528055 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30528055 Supplier Novus Biologicals Supplier No. NBP344175V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HE4/WFDC2 Monoclonal antibody specifically detects HE4/WFDC2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HE4/WFDC2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL12917
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q14508, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10406
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.