missing translation for 'onlineSavingsMsg'
Learn More

RBCK1 Antibody (CL4290) [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30528149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30528149 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30528149 Supplier Novus Biologicals Supplier No. NBP344179AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBCK1 Monoclonal antibody specifically detects RBCK1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBCK1
Applications Western Blot
Classification Monoclonal
Clone CL4290
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q9BYM8, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10616
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.