missing translation for 'onlineSavingsMsg'
Learn More

IFI27 Antibody (CL14627) [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30531661 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30531661 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30531661 Supplier Novus Biologicals Supplier No. NBP344299AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFI27 Monoclonal antibody specifically detects IFI27 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI27
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14627
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P40305, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.