Learn More
CPC Colder Products Company AseptiQuik L Connector 1in Hose Barb
Brand:  CPC Colder Products Company AQL17016
AseptiQuik L Connector 1in Hose Barb
Product Code. 30968762
  • ONLINE EXCLUSIVE PRICE
    € 122.00
    € 119.96 / Pack of 1
    Save € 2.04
    1% Off
Estimated Shipment: 15-09-2026