Description
The 3500 SERIES disposable respirators offer an outstanding level of protection demonstrated when conducting Face Fit Testing. Having electrostatic filter media allows low breathing resistance and a lightweight outer shell maintains excellent shape.
- Lightweight, cup shaped respirator
- Four point headstrap adjustment
- Preformed nosebridge removes the need for regular adjustment
- Electrostatic filter media
- Soft touch elastic
- PVC Foam Inner Face Seal
- Conforms to EN 149:2001+A1:2009 FFP3 NR
- Low Profile Exhalation Valve helps reduce heat and moisture build up
Specifications
Specifications
| Aerosol Type | R - Somewhat Resistant to Oil |
| Assigned Protection Factor (APF) | 20 |
| Certifications/Compliance | CE III , UKCA , UK Medical Device Directive: 93/42/EEC and 2007/47/EC , EU Medical Device Regulation: 2017/745 , EN 149:2000 + A1:2009 FFP3 NR |
| Color | White |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| Show More |
Product Title
Individuate un'opportunità di miglioramento?