Learn More
Corning™ 384 Well Flat Clear Bottom Polystyrene Untreated Microplates, low flange, no lid, nonsterile
Shop All Corning Products
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Description
- Recommended working volume of up to 20 to 80μL
- Conforms to standard microplate footprint and dimensions
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng lgG/cm²
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Square, Flat Bottom (F-well) |