Learn More
(3aR-cis)-Hexahydro-2-oxo-4H-furo[3,2-b]pyrrole-4-acetic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (3aR-cis)-Hexahydro-2-oxo-4H-furo[3,2-b]pyrrole-4-acetic Acid Ethyl Ester |
| CAS | 87332-79-4 |
| Molecular Formula | C10H15NO4 |
| Quantity | 25 mg |
| Synonym | Ethyl 2-((6aR)-2-Oxotetrahydro-2H-furo[3,2-b]pyrrol-4(5H)-yl)acetate |
| InChI Formula | InChI=1S/C10H15NO4/c1-2-14-10(13)6-11-4-3-8-7(11)5-9(12)15-8/h7-8H,2-6H2,1H3/t7?,8-/m1/s1 |
| SMILES | CCOC(=O)CN1CC[C@H]2OC(=O)CC12 |
| IUPAC Name | ethyl 2-[(6aR)-2-oxo-3a,5,6,6a-tetrahydro-3H-furo[3,2-b]pyrrol-4-yl]acetate |
| Molecular Weight (g/mol) | 213.23 |
| Formula Weight | 213.1001 |