Learn More
Uvex™ 3B Chem Classic Chemical Protection Coveralls
Product Code. p-7228504 Shop All Uvex Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 17797649
Brand: Uvex™ 8988009
Disposable coveralls constructed of polypropylene SMS laminated with polyethylene film for maximum safety.
  • Highly durable material combined with ultrasonically welded and taped seams ensure an effective barrier and maximum safety
  • Impervious to liquids
  • Low-noise material
  • Free of silicone and paint wetting impairment substances
  • Material: Polypropylene non woven laminated with Polypropylene film
  • Offers protection against a wide range of chemicals
  • Middle finger loops prevent sleeves sliding up the arm
  • Self-adhesive zipper flap for optimal protection
  • Anti-static properties
  • Protection against infectious diseases
  • Lightweight and durable material
  • Skin-friendly textile grip on inside
  • Elasticated waistband for a perfect fit
  • Secure and convenient closures thanks to elasticated bands on hood, arms and legs
  • Comfortable zip puller
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Polypropylene
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance EN 14126, EN 14605 - type 3B, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 1149-5, EN 1073-2