Learn More
(3Beta,5Alpha,16Beta)-3,17-Dihydroxy-16-methylpregnan-20-one, TRC
Specifications
Specifications
| Chemical Name or Material | (3beta,5alpha,16beta)-3,17-Dihydroxy-16-methylpregnan-20-one |
| CAS | 2543-24-0 |
| Molecular Formula | C22H36O3 |
| Quantity | 50 mg |
| Synonym | 3beta,17-Dihydroxy-16beta-methyl-5alpha-pregnan-20-one |
| InChI Formula | InChI=1S/C22H36O3/c1-13-11-19-17-6-5-15-12-16(24)7-9-20(15,3)18(17)8-10-21(19,4)22(13,25)14(2)23/h13,15-19,24-25H,5-12H2,1-4H3/t13-,15-,16-,17+,18-,19-,20-,21-,22-/m0/s1 |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]2(C)[C@@]1(O)C(=O)C |
| IUPAC Name | 1-[(3S,5S,8R,9S,10S,13S,14S,16S,17R)-3,17-dihydroxy-10,13,16-trimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 348.52 |
| Formula Weight | 348.2664 |