missing translation for 'onlineSavingsMsg'
Learn More

(3Beta,5Beta,7Alpha,12Alpha,24E)-3,7,12-Trihydroxy-cholest-24-en-26-oic Acid, TRC

Product Code. 30450558
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450558 2.5 mg 2.5mg
30449844 10 mg 10mg
30456008 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450558 Supplier Toronto Research Chemicals Supplier No. TRCT8879952.5MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3Beta,5Beta,7Alpha,12Alpha,24E)-3,7,12-Trihydroxy-cholest-24-en-26-oic Acid
Analyte or Component Names (3Beta,5Beta,7Alpha,12Alpha,24E)-3,7,12-Trihydroxy-cholest-24-en-26-oic Acid
Shipping Condition Room Temperature
Molecular Formula C27H44O5
InChI Formula InChI=1S/C27H44O5/c1-15(6-5-7-16(2)25(31)32)19-8-9-20-24-21(14-23(30)27(19,20)4)26(3)11-10-18(28)12-17(26)13-22(24)29/h7,15,17-24,28-30H,5-6,8-14H2,1-4H3,(H,31,32)/b16-7+/t15-,17+,18+,19-,20?,21?,22-,23+,24?,26+,27-/m1/s1
Solution Type Neat
SMILES C[C@H](CC/C=C(C)/C(O)=O)[C@@H](CC1)[C@]2(C)C1C3[C@H](O)C[C@]4([H])C[C@@H](O)CC[C@]4(C)C3C[C@@H]2O
IUPAC Name (6R,E)-2-methyl-6-((3S,5S,7R,10S,12S,13R,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)hept-2-enoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 448.64
Quantity 2.5 mg
Formula Weight 448.32
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.