Learn More
Thermo Scientific™ 3M™ Comply™ Indicator Tape
Product Code. 10514885
Click to view available options
Dimensions (L x W):
55 m x 18 mm
55 m x 19 mm
55 m x 24 mm
Unit Size:
Each
Product Code. 10514885
Brand: Thermo Scientific™ 7000136721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.
  • Lead-free steam indicator tape is designed for use on all wraps
  • Reliable achesive adheres to packaging material
  • Sustainable, solvent-free manufacturing proces
  • Does not require hazardous waste disposal
  • Redesigned packaging uses up to 30% less plastic
  • Convenient, individually wrapped, easy-to-open rolls
  • Lead free and no added latex
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets NF EN 867-1 Class A, ISO 11140-1 Class 1
Color Beige
Dimensions (L x W) 55 m x 19 mm
For Use With (Application) Sterilization verification
For Use With (Equipment) Autoclave
Length (Metric) 55 m
Product Type Indicator Tape
Features Lead-free, aggressive, pressure sensitive adhesive
Quantity 24 Pk.
Sterilization Indicator Yes
Sterilization Method Steam
Width (Metric) Exterior 19 mm