Learn More
3M™ Spectacle Kits for 7000 Series Full Facepiece Respirators
Description
- Permits the wearer to use corrective lenses and to make position adjustment that is most comfortable.
- 3M Catalog# 7925/7925-AM
Specifications
Specifications
| Product Type | Spectacle Kit |
| For Use With (Application) | Demolition, facility maintenance, grinding, sanding, welding |
| For Use With (Equipment) | Full facepiece 7000 series |
| Model | 7925/7925-AM |