Learn More
(3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone ((3'R)-Ezetimibe), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone ((3'R)-Ezetimibe)
Specifications
Specifications
| Chemical Name or Material | (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone |
| CAS | 163380-16-3 |
| Analyte or Component Names | (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H21 F2 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3R,4S)-1-(4-Fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-2-azetidinone, 3-TMR,3R,4S-Isomer of Ezetimibe, R,R,S-Ezetimibe, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-, [3R-[3alpha(R*),4beta]]-, 3’R,3R,4S-Isomer of Ezetimibe |
| InChI Formula | InChI=1S/C24H21F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21-23,28-29H,13-14H2/t21-,22-,23-/m1/s1 |
| SMILES | O[C@H](CC[C@@H]1[C@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccc(F)cc4 |
| IUPAC Name | (3R,4S)-1-(4-fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one |
| Show More |