Learn More
(3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)dihydrofuran-2(3H)-one, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 5336-08-3 |
| Molecular Formula | C5H8O5 |
| MDL Number | MFCD00063241 |
| Quantity | 25 g |
| SMILES | O[C@H]([C@@H]([C@H]1CO)O)C(O1)=O |
| Molecular Weight (g/mol) | 148.11 |
| Percent Purity | 95% |
| Chemical Name or Material | (3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)dihydrofuran-2(3H)-one |