Learn More
(3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one, Mikromol™
Product Code. 30050457
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050457
Brand: Mikromol MM0505.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one
CAS 217636-47-0
Analyte or Component Names (3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one
Shipping Condition Room Temperature
Molecular Formula C17 H15 Cl O3
Certifications/Compliance ISO 17025
Synonym Fenofibrate Imp. C (EP), (3RS)-3-[4-(4-Chlorobenzoyl)phenoxy]butan-2-one, Fenofibrate Impurity C
InChI Formula InChI=1S/C17H15ClO3/c1-11(19)12(2)21-16-9-5-14(6-10-16)17(20)13-3-7-15(18)8-4-13/h3-10,12H,1-2H3
SMILES CC(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)C
IUPAC Name 3-[4-(4-chlorobenzoyl)phenoxy]butan-2-one
Molecular Weight (g/mol) 302.75
Quantity 100 mg
Formula Weight 302.071 g/mol