missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-3-Acetamidopyrrolidine, 98%

Product Code. 11307806
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307806 5 g 5g
11397796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307806 Supplier Thermo Scientific Alfa Aesar Supplier No. L12927.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 114636-31-6 | C6H12N2O | 128.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114636-31-6
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00082748
InChI Key HDCCJUCOIKLZNM-UHFFFAOYNA-N
Synonym 3s---3-acetamidopyrrolidine, s-n-pyrrolidin-3-yl acetamide, n-3s-pyrrolidin-3-yl acetamide, s-3-acetamidopyrrolidine, acetamide, n-3s-3-pyrrolidinyl, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, s-3-acetamido-pyrrolidine, 3s-3-acetamidopyrrolidine, s-n-pyrrolidin-3-ylacetamide
PubChem CID 7021471
IUPAC Name N-[(3S)-pyrrolidin-3-yl]acetamide
SMILES CC(=O)NC1CCNC1

Specifications

Melting Point 52°C to 57°C
Boiling Point 70°C to 72°C (40 mmHg)
Quantity 5 g
UN Number UN3259
Beilstein 4904139
Optical Rotation −46.5° (c=1 in Ethanol)
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material (3S)-(-)-3-Acetamidopyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.