Learn More
4-([1,1'-Biphenyl]-4-yl)-4-oxobutanoic acid, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 36330-85-5 |
| Molecular Formula | C16H14O3 |
| MDL Number | MFCD00056701 |
| Quantity | 100 g |
| SMILES | O=C(O)CCC(C1=CC=C(C2=CC=CC=C2)C=C1)=O |
| Molecular Weight (g/mol) | 254.28 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-([1,1'-Biphenyl]-4-yl)-4-oxobutanoic acid |