Learn More
4-1BB/TNFRSF9/CD137 Antibody (2356C) [Allophycocyanin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB8382A
Product Code. 18633474
  • € 623.00 / 0.10mL
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Flow Cytometry, Agonist Activity, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-1BB/TNFRSF9/CD137
Monoclonal
APC
PBS
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Agonist Activity, CyTOF
2356C
Flow Cytometry, Agonist Activity, CyTOF-ready
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Chinese Hamster Ovary cell line CHO-derived human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011
0.1 mL
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
3604
Store at 4°C in the dark.