missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody (LOB12), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-63415C
This item is not returnable.
View return policy
Description
4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| 4-1BB/TNFRSF9/CD137 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TNFRSF9 | |
| Murine CD137 human Fc fusion protein | |
| 0.125 mL | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
| 3604 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| LOB12 | |
| Flow Cytometry | |
| 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-63415 recognises murine CD137 (the murine homologue of human 4-1BB protein). CD137 is expressed by activated T lymphocytes, but not by resting cells. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction