Learn More
4-(1H-1,2,4-Triazol-1-yl)benzoic acid, 95%, Thermo Scientific™
Product Code. 10487802
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Product Code. 10487802
Brand: Thermo Scientific Maybridge CC18601CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 162848-16-0
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.174
MDL Number MFCD02682060
InChI Key FOMQQGKCPYKKHQ-UHFFFAOYSA-N
PubChem CID 736531
IUPAC Name 4-(1,2,4-triazol-1-yl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=NC=N2
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-1h-1,2,4-triazol-1-yl benzoic acid, 4-1,2,4 triazol-1-yl-benzoic acid, 4-1,2,4-triazol-1-yl benzoic acid, benzoic acid, 4-1h-1,2,4-triazol-1-yl, benzoic acid,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzoic acid, pubchem19582, rarechem al be 1325, buttpark 9550-80, acmc-1c5ri
Formula Weight 189.17
Percent Purity 95%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzoic acid