Learn More
4-(2,6-Dichloropyrimidin-4-yl)-morpholine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2,6-Dichloropyrimidin-4-yl)-morpholine |
| CAS | 52127-83-0 |
| Molecular Formula | C8H9Cl2N3O |
| Quantity | 250 mg |
| Synonym | 2,4-Dichloro-6-(morpholin-4-yl)pyrimidine, 2,4-Dichloro-6-morpholinopyrimidine, 6-Morpholino-2,4-dichloropyrimidine |
| InChI Formula | InChI=1S/C8H9Cl2N3O/c9-6-5-7(12-8(10)11-6)13-1-3-14-4-2-13/h5H,1-4H2 |
| SMILES | Clc1cc(nc(Cl)n1)N2CCOCC2 |
| IUPAC Name | 4-(2,6-dichloropyrimidin-4-yl)morpholine |
| Molecular Weight (g/mol) | 234.08 |
| Formula Weight | 233.0123 |