Learn More
4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino]benzoic Acid (Pteroic Acid), Mikromol™
Description
4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino]benzoic Acid (Pteroic Acid)
Specifications
Specifications
| Chemical Name or Material | Pteroic acid |
| CAS | 119-24-4 |
| Analyte or Component Names | Pteroic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H12 N6 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino]benzoic Acid, Folic Acid Imp. D (EP), Pteroic Acid, Folic Acid Hydrate Impurity D, Folic Acid Impurity D |
| InChI Formula | InChI=1S/C14H12N6O3/c15-14-19-11-10(12(21)20-14)18-9(6-17-11)5-16-8-3-1-7(2-4-8)13(22)23/h1-4,6,16H,5H2,(H,22,23)(H3,15,17,19,20,21) |
| SMILES | NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)O)cnc2N1 |
| IUPAC Name | 4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoic acid |
| Show More |