Learn More
4-(2-Aminophenyl)-3-morpholinone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2-Aminophenyl)-3-morpholinone |
| CAS | 1082588-73-5 |
| Molecular Formula | C10 H12 N2 O2 |
| Quantity | 100 mg |
| Synonym | 4-(2-Aminophenyl)morpholin-3-one |
| InChI Formula | InChI=1S/C10H12N2O2/c11-8-3-1-2-4-9(8)12-5-6-14-7-10(12)13/h1-4H,5-7,11H2 |
| SMILES | Nc1ccccc1N2CCOCC2=O |
| IUPAC Name | 4-(2-aminophenyl)morpholin-3-one |
| Molecular Weight (g/mol) | 192.214 |
| Formula Weight | 192.0899 |