missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Cyano-2-propyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15469937
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15469937

Brand: Thermo Scientific Alfa Aesar H53173.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be
TRUSTED_SUSTAINABILITY

Specifications

CAS 850568-67-1
Melting Point 146°C to 148°C
Molecular Formula C10H12BNO2
MDL Number MFCD06659832
Quantity 250 mg
Synonym 2-4-boronophenyl-2-methylpropanenitrile, 4-2-cyanopropan-2-yl phenylboronic acid, 4-1-cyano-1-methylethyl phenyl boronic acid, 2-4-boronophenyl-2-methylpropionitrile, 4-2-cyanopropan-2-yl benzeneboronic acid, 4-2-cyanopropan-2-yl phenyl boronic acid, 4-1-cyano-1-methylethyl phenylboronic acid, 4-2-cyanoprop-2-yl benzeneboronic acid, boronic acid, 4-1-cyano-1-methylethyl phenyl
Solubility Information Soluble in water. [9g/100 mL (20°C)]
InChI Key BNXBFDTWYHAEIR-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(C)(C)C#N)(O)O
IUPAC Name [4-(2-cyanopropan-2-yl)phenyl]boronic acid
Molecular Weight (g/mol) 189.021
PubChem CID 44119324
Formula Weight 189.02
Percent Purity 97%
Chemical Name or Material 4-(2-Cyano-2-propyl)benzeneboronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.