missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. p-7035438
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15410436

Brand: Thermo Scientific Alfa Aesar H53155.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-34-3
Molecular Formula C10H14BNO4
Molecular Weight (g/mol) 223.035
MDL Number MFCD04115682
InChI Key QRGVHOKYRIQQTN-UHFFFAOYSA-N
Synonym 4-2-methoxyethylcarbamoyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl phenylboronic acid, 4-2-methoxyethylaminocarbonyl benzeneboronic acid, 4-2-methoxyethyl carbamoyl phenylboronic acid, acmc-209q2x, 4-2-methoxyethyl carbamoyl phenyl boronic acid, 4-2-methoxyethyl aminocarbonyl benzeneboronic acid, boronic acid,b-4-2-methoxyethyl amino carbonyl phenyl, boronicacid,b-4-2-methoxyethyl amino carbonyl phenyl
PubChem CID 44119550
IUPAC Name [4-(2-methoxyethylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NCCOC)(O)O

Specifications

Melting Point 120°C to 124°C
Quantity 250 mg
Formula Weight 223.04
Percent Purity 98%
Chemical Name or Material 4-(2-Methoxyethylcarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt