Learn More
Memmert™ 4-20 mA Current Loop Interface for IF55, Actual Value
Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Description
- Temperature controller actual value
Specifications
Specifications
| For Use With (Equipment) | IF55 Incubator |
| Type | Current Loop Interface |
| Amperage | 4 to 20 mA |