Learn More
Memmert™ 4-20 mA Current Loop Interface for IF55plus, Actual Value
Product Code. 17557145 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF55plus Incubator
Unit Size:
Each
Product Code. 17557145
missing translation for 'mfr': Memmert™ IF55plusV3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) IF55plus Incubator
Type Current Loop Interface
Amperage 4 to 20 mA