Learn More
Memmert™ 4-20 mA Current Loop Interface for IN160, Actual Value
Product Code. 17537525 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 17537525
Brand: Memmert™ IN160V3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) IN160 Incubator
Type Current Loop Interface
Amperage 4 to 20 mA