Learn More
Memmert™ 4-20 mA Current Loop Interface for UF30plus
Product Code. 17519315 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 17519315
Brand: Memmert™ UF30plusV6
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
For Use With (Equipment) UF30plus Oven
Type Current Loop Interface
Amperage 4 to 20 mA