missing translation for 'onlineSavingsMsg'
Learn More

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Catalog Number p-3586740
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
56935-79-6
C12H11N3
197.241
MFCD09817559
BTXLPDNFEZIQBZ-UHFFFAOYSA-N
4-3,5-dimethyl-1h-pyrazol-1-yl benzonitrile, 4-3,5-dimethylpyrazol-1-yl benzonitrile, acmc-209luz, 4-3,5-dimethylpyrazol-1-yl-benzonitrile, 4-3,5-dimethylpyrazolyl benzenecarbonitrile, 1-4-cyanophenyl-3,5-dimethyl-1h-pyrazole, benzonitrile, 4-3,5-dimethyl-1h-pyrazol-1-yl
15005387
4-(3,5-dimethylpyrazol-1-yl)benzonitrile
CC1=CC(=NN1C2=CC=C(C=C2)C#N)C
This item is not returnable. View return policy
56935-79-6
C12H11N3
197.241
MFCD09817559
BTXLPDNFEZIQBZ-UHFFFAOYSA-N
4-3,5-dimethyl-1h-pyrazol-1-yl benzonitrile, 4-3,5-dimethylpyrazol-1-yl benzonitrile, acmc-209luz, 4-3,5-dimethylpyrazol-1-yl-benzonitrile, 4-3,5-dimethylpyrazolyl benzenecarbonitrile, 1-4-cyanophenyl-3,5-dimethyl-1h-pyrazole, benzonitrile, 4-3,5-dimethyl-1h-pyrazol-1-yl
15005387
4-(3,5-dimethylpyrazol-1-yl)benzonitrile
CC1=CC(=NN1C2=CC=C(C=C2)C#N)C
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.24
Percent Purity 97%
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'