Learn More
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile, 97%, Thermo Scientific™
Product Code. 15463256
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463256 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15463256 Supplier Thermo Scientific Alfa Aesar Supplier No. H57257.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1421262-65-8
Color White
Molecular Formula C11H9N3O
MDL Number MFCD22682905
Quantity 250 mg
UN Number UN3439
Synonym 4-3-ethyl-1,2,4-oxadiazol-5-yl benzonitrile
InChI Key OLEJLLUUIDNJDM-UHFFFAOYSA-N
SMILES CCC1=NOC(=N1)C2=CC=C(C=C2)C#N
IUPAC Name 4-(3-ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
Molecular Weight (g/mol) 199.213
PubChem CID 73996265
Formula Weight 199.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile