Learn More
4-[3-(Methylsulfonyl)phenyl]-1-propyl-1,2,3,6-tetrahydropyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[3-(Methylsulfonyl)phenyl]-1-propyl-1,2,3,6-tetrahydropyri |
| CAS | 346688-39-9 |
| Molecular Formula | C15 H21 N O2 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C15H21NO2S/c1-3-9-16-10-7-13(8-11-16)14-5-4-6-15(12-14)19(2,17)18/h4-7,12H,3,8-11H2,1-2H3 |
| SMILES | CCCN1CCC(=CC1)c2cccc(c2)S(=O)(=O)C |
| IUPAC Name | 4-(3-methylsulfonylphenyl)-1-propyl-3,6-dihydro-2H-pyridine |
| Molecular Weight (g/mol) | 279.4 |
| Formula Weight | 279.1293 |