missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(3-Methylsulfonylphenyl)-1-propylpiperidine, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | 4-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine |
| CAS | 346688-38-8 |
| Molecular Formula | C15 H23 N O2 S |
| Quantity | 1 mg |
| Synonym | Pridopidine |
| InChI Formula | InChI=1S/C15H23NO2S/c1-3-9-16-10-7-13(8-11-16)14-5-4-6-15(12-14)19(2,17)18/h4-6,12-13H,3,7-11H2,1-2H3 |
| SMILES | CCCN1CCC(CC1)c2cccc(c2)S(=O)(=O)C |
| IUPAC Name | 4-(3-methylsulfonylphenyl)-1-propylpiperidine |
| Molecular Weight (g/mol) | 281.41 |
| Formula Weight | 281.1449 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering